SY3H3D67C32F Synthonix Corporation

3,4-Dimethoxynitrobenzene

  • MDL number MFCD00007238
  • Cas Number : 709-09-1
  • Molecular Weight : 183.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,2-dimethoxy-4-nitrobenzene
Inchi Code 1S/C8H9NO4/c1-12-7-4-3-6(9(10)11)5-8(7)13-2/h3-5H,1-2H3
Inchi Key YFWBUVZWCBFSQN-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/49347.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338,P310
MSDS https://synthonix.com/49347.html
pictograms GHS07
hstatements H302,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart