SY3H3D67C33F Synthonix Corporation

(2,6-Difluoro-3,5-dimethoxyphenyl)hydrazine hydrochloride

  • Cas Number : 1841081-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49503.html

Safety Information

MSDS https://synthonix.com/49503.html
Availability details for
Quantity
  • Availability
  • Add to Cart