SY3H3D67C340 Synthonix Corporation

4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)butanehydrazide hydrochloride

  • MDL number MFCD22372692
  • Cas Number : 1588441-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49519.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49519.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart