SY3H3D67C346 Synthonix Corporation

3-(N,N-Dimethylamino)phenylboronic acid hydrochloride

  • MDL number MFCD04112542
  • Cas Number : 1256355-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49543.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49543.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart