SY3H3D67C3C3 Synthonix Corporation

N,N-Dimethyl-1H-indazol-5-amine

  • Cas Number : 945264-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49996.html

Safety Information

MSDS https://synthonix.com/49996.html
Availability details for
Quantity
  • Availability
  • Add to Cart