SY3H3D67C3C4 Synthonix Corporation

1,5-Dimethyl-1H-benzo[d]imidazol-2-amine

  • Cas Number : 39860-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49998.html

Safety Information

MSDS https://synthonix.com/49998.html
Availability details for
Quantity
  • Availability
  • Add to Cart