SY3H3D67C3C7 Synthonix Corporation

1-(3,5-Dichloropyridin-4-yl)piperazine

  • MDL number MFCD00221400
  • Cas Number : 175277-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White - Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50003.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50003.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart