SY3H3D67C3C8 Synthonix Corporation

1,3-Diallyl-1,3,5-triazinane-2,4,6-trione

  • MDL number MFCD00767871
  • Cas Number : 6294-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - blue
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50011.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50011.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart