SY3H3D67C3D5 Synthonix Corporation

N,N-Dimethyl-1-(4-nitrophenyl)methanamine

  • MDL number MFCD00035956
  • Cas Number : 15184-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50186.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50186.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart