SY3H3D67C3DB Synthonix Corporation

(2,4-Dimethylphenyl)methanol

  • MDL number MFCD00004624
  • Cas Number : 16308-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Lump / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50273.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50273.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart