SY3H3D67C3F3 Synthonix Corporation

Diethyl 2-methyl-3-oxosuccinate

  • MDL number MFCD00009163
  • Cas Number : 759-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Clear liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50455.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50455.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart