SY3H3D67C3FD Synthonix Corporation

Diethyl 2-hydroxypentanedioate

  • MDL number MFCD16620610
  • Cas Number : 69134-53-8
  • Molecular Weight : 204.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50526.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50526.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart