SY3H3D67C418 Synthonix Corporation

1,4-Dichloroisoquinoline

  • Cas Number : 15298-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50733.html

Safety Information

MSDS https://synthonix.com/50733.html
Availability details for
Quantity
  • Availability
  • Add to Cart