SY3H3D67C426 Synthonix Corporation

4-(3,4-Dichlorophenyl)thiazol-2-amine

  • MDL number MFCD00466323
  • Cas Number : 39893-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50805.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50805.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart