SY3H3D67C42A Synthonix Corporation

2,3-Dichloro-6-methoxyquinoxaline

  • MDL number MFCD00461063
  • Cas Number : 39267-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white to gray powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50838.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50838.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart