SY3H3D67C439 Synthonix Corporation

1,4-Difluoro-2-(prop-2-yn-1-yloxy)benzene

  • Cas Number : 457628-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51032.html

Safety Information

MSDS https://synthonix.com/51032.html
Availability details for
Quantity
  • Availability
  • Add to Cart