SY3H3D67C52A Synthonix Corporation

1-(4-(Difluoromethoxy)phenyl)ethanone

  • MDL number MFCD00042250
  • Cas Number : 83882-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless to Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51616.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51616.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart