SY3H3D67C54D Synthonix Corporation

2,3-Dihydrobenzofuran-5-carboxamide

  • Cas Number : 262847-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51783.html

Safety Information

MSDS https://synthonix.com/51783.html
Availability details for
Quantity
  • Availability
  • Add to Cart