SY3H3D67C54E Synthonix Corporation

2,4-Dichloro-D-phenylalanine

  • MDL number MFCD01860873
  • Cas Number : 114872-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51795.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51795.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart