SY3H3D67C552 Synthonix Corporation

2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde oxime

  • MDL number MFCD00862517
  • Cas Number : 31127-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51826.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51826.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart