SY3H3D67C55E Synthonix Corporation

(S)-1-(2,5-Difluorophenyl)propan-1-amine hydrochloride

  • Cas Number : 1391431-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52098.html

Safety Information

MSDS https://synthonix.com/52098.html
Availability details for
Quantity
  • Availability
  • Add to Cart