SY3H3D67C564 Synthonix Corporation

4,7-Difluoro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD30609572
  • Cas Number : 2061980-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52229.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52229.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart