SY3H3D67C565 Synthonix Corporation

5,7-Difluoro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • Cas Number : 1229784-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52230.html

Safety Information

MSDS https://synthonix.com/52230.html
Availability details for
Quantity
  • Availability
  • Add to Cart