SY3H3D67C56B Synthonix Corporation

(R)-1-(2,2-Difluorobenzo[d][1,3]dioxol-4-yl)ethanamine hydrochloride

  • Cas Number : 1257106-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52241.html

Safety Information

MSDS https://synthonix.com/52241.html
Availability details for
Quantity
  • Availability
  • Add to Cart