SY3H3D67C575 Synthonix Corporation

3-(Dimethylamino)-2-(thiophene-2-carbonyl)acrylonitrile

  • MDL number MFCD00120337
  • Cas Number : 52200-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52339.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52339.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart