SY3H3D67C577 Synthonix Corporation

N,N-Dimethyl-2-(perfluorophenoxy)ethanamine

  • MDL number MFCD30478399
  • Cas Number : 2044704-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52404.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52404.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart