SY3H3D67C5CC Synthonix Corporation

3,4-Dihydro-2H-benzo[b][1,4]dioxepine-2-carboxylic acid

  • MDL number MFCD09953183
  • Cas Number : 33632-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52490.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52490.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart