SY3H3D67C5D5 Synthonix Corporation

3-(3,4-Dichlorophenyl)pyrrolidine hydrochloride

  • Cas Number : 1092108-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52537.html

Safety Information

MSDS https://synthonix.com/52537.html
Availability details for
Quantity
  • Availability
  • Add to Cart