SY3H3D67C5DB Synthonix Corporation

4,6-Dimethoxyindolin-2-one

  • MDL number MFCD11845488
  • Cas Number : 23659-88-3
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52586.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52586.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart