SY3H3D67C5E0 Synthonix Corporation

Dimethyl (3-oxo-1,3-dihydroisobenzofuran-1-yl)phosphonate

  • MDL number MFCD16883476
  • Cas Number : 61260-15-9
  • Molecular Weight : 242.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52664.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52664.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart