SY3H3D67C5E1 Synthonix Corporation

1,2-Dibromo-4-tert-butylbenzene

  • MDL number MFCD05664303
  • Cas Number : 6683-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow Solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52676.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52676.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart