SY3H3D67C5EA Synthonix Corporation

N,N-Dimethyl-2-(3,4,5-trifluorophenoxy)ethanamine

  • Cas Number : 1708924-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52776.html

Safety Information

MSDS https://synthonix.com/52776.html
Availability details for
Quantity
  • Availability
  • Add to Cart