SY3H3D67C5ED Synthonix Corporation

1-(3,5-Dimethylphenyl)ethanone

  • MDL number MFCD01075693
  • Cas Number : 5379-16-8
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-dimethylphenyl)ethanone
Inchi Code 1S/C10H12O/c1-7-4-8(2)6-10(5-7)9(3)11/h4-6H,1-3H3
Inchi Key BKIHFZLJJUNKMZ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Liquid - Crystal - Lump
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/52785.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/52785.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart