SY3H3D67C5EF Synthonix Corporation

2,4-Dimethylphenylacetic Acid

  • MDL number MFCD02664685
  • Cas Number : 6331-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52798.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52798.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart