SY3H3D67C5FD Synthonix Corporation

Diethyl furan-2,5-dicarboxylate

  • MDL number MFCD00449457
  • Cas Number : 53662-83-2
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52861.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52861.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart