SY3H3D67C60D Synthonix Corporation

N,N-Diethyl-m-nitroaniline

  • Cas Number : 2216-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53045.html

Safety Information

MSDS https://synthonix.com/53045.html
Availability details for
Quantity
  • Availability
  • Add to Cart