SY3H3D67C619 Synthonix Corporation

2,2-Dichloro-1-(1-oxa-4-azaspiro[4.5]decan-4-yl)ethanone

  • MDL number MFCD00184232
  • Cas Number : 71526-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53156.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53156.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart