SY3H3D67C61A Synthonix Corporation

(3aR,6S)-8,8-Dimethyl-4,5,6,7-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide

  • MDL number MFCD09992877
  • Cas Number : 107869-45-4
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53160.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53160.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart