SY3H3D67C61B Synthonix Corporation

(3aS,6R)-8,8-Dimethyl-4,5,6,7-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide

  • MDL number MFCD09992876
  • Cas Number : 60886-80-8
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3aS,6R)-8,8-dimethyl-1,4,5,6-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide
Inchi Code 1S/C10H15NO2S/c1-9(2)7-3-4-10(9)6-14(12,13)11-8(10)5-7/h5,7,11H,3-4,6H2,1-2H3/t7-,10-/m1/s1
Inchi Key LDJJQEAMNCWNKP-GMSGAONNSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/53161.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/53161.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart