SY3H3D67C621 Synthonix Corporation

dimethyl (1S,2R)-cyclohexane-1,2-dicarboxylate

  • MDL number MFCD00043768
  • Cas Number : 1687-29-2
  • Molecular Weight : 200.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name dimethyl (1R,2S)-1,2-cyclohexanedicarboxylate
Inchi Code 1S/C10H16O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h7-8H,3-6H2,1-2H3/t7-,8+
Inchi Key AIACXWOETVLBIA-OCAPTIKFSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Colorless - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/53215.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/53215.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart