SY3H3D67C626 Synthonix Corporation

(3aS,6S,7aS)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide

  • Cas Number : 94594-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53232.html

Safety Information

MSDS https://synthonix.com/53232.html
Availability details for
Quantity
  • Availability
  • Add to Cart