SY3H3D67C62B Synthonix Corporation

Diethyl 3,3'-thiodipropanoate

  • MDL number MFCD00026920
  • Cas Number : 673-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53280.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53280.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart