SY3H3D67C633 Synthonix Corporation

N,N-Dibutylacetamide

  • MDL number MFCD00027190
  • Cas Number : 1563-90-2
  • Molecular Weight : 171.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53344.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53344.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart