SY3H3D67C642 Synthonix Corporation

4,6-Dichloro-5-phenylpyrimidine

  • Cas Number : 3974-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53428.html

Safety Information

MSDS https://synthonix.com/53428.html
Availability details for
Quantity
  • Availability
  • Add to Cart