SY3H3D67C643 Synthonix Corporation

4,4'-Difluoro-2,2'-bipyridine

  • Cas Number : 1189458-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53429.html

Safety Information

MSDS https://synthonix.com/53429.html
Availability details for
Quantity
  • Availability
  • Add to Cart