SY3H3D67C650 Synthonix Corporation

Di(pyridin-2-yl) sulfite

  • MDL number MFCD27991313
  • Cas Number : 105125-43-7
  • Molecular Weight : 236.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name di(pyridin-2-yl) sulfite
Inchi Code 1S/C10H8N2O3S/c13-16(14-9-5-1-3-7-11-9)15-10-6-2-4-8-12-10/h1-8H
Inchi Key YIZMNGKCMZDQJW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/53679.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/53679.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart