SY3H3D67C65B Synthonix Corporation

5,7-Dimethylindoline-2,3-dione

  • MDL number MFCD00047219
  • Cas Number : 39603-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53926.html

Safety Information

signalword Warning
pcodes P273
MSDS https://synthonix.com/53926.html
pictograms GHS07
hstatements H302, H319, H412
Availability details for
Quantity
  • Availability
  • Add to Cart