SY3H3D67C6C5 Synthonix Corporation

1,3-Di(pyridin-2-yl)urea

  • MDL number MFCD00233705
  • Cas Number : 6268-43-5
  • Molecular Weight : 214.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54306.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54306.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart