SY3H3D67C6C8 Synthonix Corporation

2,8-Dimethylquinoline

  • Cas Number : 1463-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54370.html

Safety Information

MSDS https://synthonix.com/54370.html
Availability details for
Quantity
  • Availability
  • Add to Cart