SY3H3D67C6CE Synthonix Corporation

3-(Dimethylamino)-1-(3-nitrophenyl)prop-2-en-1-one

  • MDL number MFCD00024513
  • Cas Number : 115955-48-1
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/54496.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/54496.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart